VWR® LABWASH® Premium Detergents and Cleaning Agents
Toimittaja: VWR Chemicals
84545.360EA 105 EUR
84545.360 84545.410 84546.360 84546.410 84547.360 84547.410 84548.290 84548.410 84549.360 84549.410 84550.360 84550.410 84550.440 84551.360 84551.410 84551.440 84552.360 84552.410 84553.360 84554.360 84555.360 84556.360 84556.410 84557.290 84557.410 84861.410 84545.320 84546.320 84547.320 84549.320 84550.320 84551.320 84553.320 84554.320 84549.365 84550.365 84551.365 84552.365 84553.365 84555.365 84556.365 85831.360
VWR® LABWASH® Premium Detergents and Cleaning Agents
Puhdistusaineet Pesuaineet ja pintojen puhdistusaineet
Compact HDPE canisters (Cat. Nos. 845xx.365) are compatible with Miele dosing cabinets.
  • Residue-free laboratory glassware cleaner
  • Excellent cleaning power at low dosage levels
  • NTA-free (no nitrilotriacetic acid)
  • Protects the environment, active ingredients are biodegradable
  • Acid-rinse concentrates available to neutralise alkaline cleaners
pH value at 1% concentration.