Metyleenikloridi, anhydrous (max. 0.001% H₂O) ≥99.8% stabilised

Toimittaja: VWR Chemicals
Varoitus

Synonyms: Methylene chloride

Stabilization: 20 ppm Stabilised with 2-methyl-2-butene (iso-amylene)
Filtered through a 0.2 µm filter, packaged under nitrogen.

83682.230EA 78.2 EUR
83682.230 83682.290
Metyleenikloridi, anhydrous (max. 0.001% H₂O) ≥99.8% stabilised
Metyleenikloridi

250 ml bottle with a septum cap featuring six separate re-sealable puncture points

Kaava: CH₂Cl₂
MW: 84.93 g/mol
Kiehumispiste: 39.8 °C (1013 hPa)
Sulamispiste: –95 °C
Tiheys: 1.322 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000881
CAS-numero: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
REACH: 01-2119480404-41
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (on anhydrous substance) Min. 99.8 %
Acidity Max. 0.0005 meq/g
Evaporation residue Max. 5 ppm
Water Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR