2-Propanoli, anhydrous (max. 0.003% H₂O) ≥99.8%

Toimittaja: VWR Chemicals
Vaara

Synonyms: Isopropyylialkoholi

Filtered through a 0.2 µm filter, packaged under nitrogen.

83677.230EA 67.8 EUR
83677.230 83677.290
2-Propanoli, anhydrous (max. 0.003% H₂O) ≥99.8%
2-Propanoli

250 ml bottle with a septum cap featuring six separate re-sealable puncture points

Kaava: (CH₃)₂CHOH
MW: 60.1 g/mol
Kiehumispiste: 82
Sulamispiste: –89 °C
Tiheys: 0.786 g/cm³ (20 °C)
Leimahduspiste: 12 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011674
CAS-numero: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (calculated on anhydrous) Min. 99.8 %
Acidity Max. 0.0005 meq/g
Evaporation residue Max. 0.0005 %
Water Max. 0.0030 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR