Metyleenikloridi 99,8 Stabilised, PESTINORM® Pest_Residual

Toimittaja: VWR Chemicals
Varoitus

Synonyms: Methylene chloride

Stabilization: 20 ppm 2-Methyl-2-buten
Filter 0,2 um_Nitrogen_filled

83665.320EA 164 EUR
83665.320 83665.290 83665.400
Metyleenikloridi 99,8 Stabilised, PESTINORM® Pest_Residual
Metyleenikloridi
Kaava: CH₂Cl₂
MW: 84.93 g/mol
Kiehumispiste: 39.8 °C (1013 hPa)
Sulamispiste: –95 °C
Tiheys: 1.322 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000881
CAS-numero: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
REACH: 01-2119480404-41
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (on anhydrous substance) Min. 99,8%
Water Max. 0,02 %
Evaporation residue Max. 0,0005 %
Pesticide analysis (Lindane/ECD) Max. 5 ng/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR