Booritrifluoridi liuos ∼10% (∼1,3 M) in 1-butanol, LiChropur™ for GC derivatisation, Supelco®

Toimittaja: Merck
Vaara


M-Clarity™ quality level = MQ100

83253-100ML-F
83253-100ML-FEA 179 EUR
83253-100ML-F
Booritrifluoridi liuos ∼10% (∼1,3 M) in 1-butanol, LiChropur™ for GC derivatisation, Supelco®
Booritrifluoridi

BF3 - Butanol solution

Kaava: BF₃
Storage Temperature: Jääkaappi
MDL numero: MFCD00011316
CAS-numero: 7637-07-2
UN: 2920
ADR: 3(8),I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9226f0e996fbdc53196b812113922992f82a16be&vid=6231819666f5cb0f086eda51954ac6cdb16128c8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR