Asetonitriili ≥99.8% (by GC), BAKER ANALYZED®, Isocratic Grade for HPLC, J.T.Baker®

Toimittaja: Avantor
Vaara

Synonyms: Syaanimetaani

8257.1000EA 89.53 EUR
8257.1000 8257.2500 8257.5000 8257.9010RC 8257.9030RC 8257-333
Asetonitriili ≥99.8% (by GC), BAKER ANALYZED®, Isocratic Grade for HPLC, J.T.Baker®
Asetonitriili
Kaava: H₃CCN
MW: 41.05 g/mol
Kiehumispiste: 81.6
Sulamispiste: –45.7 °C
Tiheys: 0.782 g/cm³ (20 °C)
Leimahduspiste: 2 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001878
CAS-numero: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (by GC) min. 99.8 %
Residue after Evaporation max. 0.0005 %
Water (H2O) max. 0.1 %
Ultraviolet Absorbance (1.00-cm path vs water):
at 200 nm max. 0.5
at 210 nm max. 0.3
at 220 nm max. 0.2
at 254 nm max. 0.05
at 280 nm max. 0.01
UV Cut-off, nm max. 190


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR