VWR®, Labelling Tapes, Wafer Box Sealing
Toimittaja: VWR Collection
817-0100EA 24.6 EUR
817-0100 817-0095 817-0059 817-0063 817-0065 817-0101 817-0096 817-0099 817-0097 817-0098 817-0152
VWR®, Labelling Tapes, Wafer Box Sealing
Teipit
These tapes, with polyethylene backing and acrylic adhesive, have low out-gassing and good clean-peel properties. They are mostly used for colour coding and wafer box sealing.
  • Over labelling (to protect paper labels)
  • Maintenance
  • Colour coding
Processed in a controlled environment, tapes are wound on a plastic core and single-bagged.
Length: 32,9 m
Different colours available.