Vanadium(III) bromide ≥99.5% (metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara

81135.04 81135.09 81135.18
81135.04EA 180 EUR
81135.04 81135.09 81135.18
Vanadium(III) bromide ≥99.5% (metals basis)
Vanadium(III) bromide

Purity: 99.5% CAS No.: 13470-26-3 Molecular Formula: VBr3 Formula Weight: 290.67 Physical Form: Crystalline Density: 4.00 Melting Point: dec. UN No.: 2923 MDL No.: MFCD00049695

Kaava: Br₃V
MW: 290.65 g/mol
Sulamispiste: Dec.
Tiheys: 4.00
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00049695
CAS-numero: 13470-26-3
EINECS: 236-736-6
UN: 3260
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR