Piperidiini ≥99.0% (by GC, titration analysis), Puriss. p.a., Riedel-de Haën™

Toimittaja: Honeywell Chemicals
Vaara

80640-1L 80640-500ML
80640-1LEA 419.01 EUR
80640-1L 80640-500ML
Piperidiini ≥99.0% (by GC, titration analysis), Puriss. p.a., Riedel-de Haën™
Piperidiini

Beilstein Number: 102438
RTECS Number: TM3500000
UNSPSC Code: 12190000

Kaava: C₅H₁₁N
Kiehumispiste: 10,3 %(V)
Sulamispiste: –10 °C
Tiheys: 0,86 g/cm³ (20 °C)
Leimahduspiste: 16 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005979
CAS-numero: 110-89-4
EINECS: 203-813-0
UN: 2401
ADR: 8,I
Merck Index: 12,07621

Order Now

Testitulosten erittely

Refractive Index n20/D 1.452 (lit.)
Identity (IR) complying
Assay (GC) Min. 99.5 %
Refractive index (n 20/D) 1.452 - 1.454
Non-volatile matter Max. 0.02 %
Water (Karl Fischer) Max. 0.3 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Tin (Sn) Max. 0.1 ppm

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR