Trifluorietikkahappo

Toimittaja: Merck

Synonyms: TFA

80457-10ML
80457-10MLEA 164 EUR
80457-10ML
Trifluorietikkahappo
Trifluorietikkahappo
Kaava: C₂HF₃O₂
MW: 114.02 g/mol
Kiehumispiste: 71.8
Sulamispiste: -15
MDL numero: MFCD00004169
CAS-numero: 76-05-1
Merck Index: 13,09754

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR