n-Heksaani, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Toimittaja: Avantor
Vaara

Synonyms: Hexyl hydride, Hexanen, Hexane

8044.1000EA 76.1 EUR
8044.1000 8044.2500 8044.9025 8044.5000 8044.9200
n-Heksaani, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
n-Heksaani
Kaava: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Kiehumispiste: 69
Sulamispiste: –94.3 °C
Tiheys: 0.659 g/cm³ (20 °C)
Leimahduspiste: –22 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009520
CAS-numero: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (by GC) min. 95 %
Aromatic Compounds (as C6H6) max. 0.02 %
Color (APHA) max. 10
Density (g/ml) at 25°C max. 0.657
Residue after Evaporation max. 0.001 %
Sulfur Compounds (as S) max. 0.005 %
Thiophene passes test
Water Soluble Titrable Acid (meq/g) max. 0.0003
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR