Etanoli absolute, BAKER ANALYZED® ACS, J.T.Baker®

Toimittaja: Avantor
Vaara

Synonyms: Ethyl alcohol

8025.1000EA 41.01 EUR
8025.1000 8025.0250 8025.9025 8025.5000PE 8025.2500PE 8025.2500 8025.1000PE 8025.9200
Etanoli absolute, BAKER ANALYZED® ACS, J.T.Baker®
Etanoli
Kaava: C₂H₅OH
Kiehumispiste: 78,3 °C (1013 hPa)
Sulamispiste: –117 °C
Tiheys: 0,7895 g/cm³ (20 °C)
Leimahduspiste: 12 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003568
CAS-numero: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
REACH: 01-2119457610-43
Merck Index: 12,03806

Order Now

Testitulosten erittely

Exceeds ACS Specifications
Assay min. 99.9 % (v/v)
Acetone, isopropylalcohol passes test
Color (APHA) max. 10
Methanol (CH3OH) max. 0.1 %
Residue after Evaporation max. 0.001 %
Solubility in Water passes test
Substances Darkened by H2SO4 passes test
Substances Reducing KMnO4 passes test
Titrable Acid (meq/g) max. 0.0005
Titrable Base (meq/g) max. 0.0002
Water (H2O) max. 0.2 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.2
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR