1-Butanoli, BAKER ANALYZED® ACS, J.T.Baker®

Toimittaja: Avantor
Vaara

Synonyms: 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol

8017.2500EA 151 EUR
8017.2500 8017.1000 8017.9025
1-Butanoli, BAKER ANALYZED® ACS, J.T.Baker®
1-Butanoli
Kaava: H₃C(CH₂)₂CH₂OH
Kiehumispiste: 117,6 °C (1013 hPa)
Sulamispiste: –89 °C
Tiheys: 0,812 g/cm³ (20 °C)
Leimahduspiste: 30 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002964
CAS-numero: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3,III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Exceeds ACS Specifications
Assay (by GC) min. 99.4 %
Aldehydes passes test
Butyl Ether (by GC) max. 0.2 %
Carbonyl Compounds (as butyraldehyde) max. 0.01 %
Color (APHA) max. 10
Residue after Evaporation max. 0.001 %
Titrable Acid (meq/g) max. 0.0008
Water (H2O) max. 0.1 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR