Acetylacetonato(1,5-cyclooctadiene)rhodium(I) for synthesis

Toimittaja: Merck
Varoitus

Synonyms: (Acetylacetonato)(1,5-cyclooctadiene)rhodium(I)

M-Clarity™ quality level = MQ200

8.24420.0100 8.24420.0500
8.24420.0100EA 186 EUR
8.24420.0100 8.24420.0500
Acetylacetonato(1,5-cyclooctadiene)rhodium(I) for synthesis
Acetylacetonato(1,5-cyclooctadiene)rhodium(I)
Kaava: C₁₃H₂₀O₂Rh
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00075046
CAS-numero: 12245-39-5

Order Now

Testitulosten erittely

Solubility Insoluble (20 °C)
Melting Point 138 - 140 °C
Molar Mass 310.2 g/mol
Melting point +138...+140 °C
Assay (to C, H) ≥96%
Identity (IR) Passes test
Storage temperature +15...+25 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR