Fenoli for synthesis, Sigma-Aldrich®

Toimittaja: Merck
Vaara

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

M-Clarity™ quality level = MQ200

8.22296.0100 8.22296.0500 8.22296.1000
8.22296.0100EA 39.5 EUR
8.22296.0100 8.22296.0500 8.22296.1000
Fenoli for synthesis, Sigma-Aldrich®
Fenoli
Kaava: C₆H₅OH
Kiehumispiste: 181,7 °C (1013 hPa)
Sulamispiste: 40,8 °C
Tiheys: 1,0722 g/cm³ (20 °C)
Leimahduspiste: 81 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002143
CAS-numero: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1,II
REACH: 01-2119471329-32
Merck Index: 12,07390

Order Now

Testitulosten erittely

Solubility 84 g/l (20 °C)
Melting Point 40.8 °C
Molar Mass 94.11 g/mol
Bulk Density 620 kg/m3
Boiling Point 181.8 °C (1013 hPa)
Vapor Pressure 0.2 hPa (20 °C)
Flash Point 81 °C
Explosion Limit 1.3 - 9.5 %(V)
Density 1.06 g/cm3 (20 °C)
pH 5 (50 g/l, H2O, 20 °C)
Ignition Point 595 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR