Asetoni, EMPLURA®, Reagent Grade for synthesis, Supelco®

Toimittaja: Merck
Vaara

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

M-Clarity™ quality level = MQ200

8.22251.1000 8.22251.2500 8.22251.9025
8.22251.1000EA 24 EUR
8.22251.1000 8.22251.2500 8.22251.9025
Asetoni, EMPLURA®, Reagent Grade for synthesis, Supelco®
Asetoni
Kaava: CH₃COCH₃
Kiehumispiste: 56,2 °C (1013 hPa)
Sulamispiste: –95,4 °C
Tiheys: 0,792 g/cm³ (20 °C)
Leimahduspiste: <–20 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008765
CAS-numero: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR