2-Ethylhexyl phosphate (mono and di ester mixture) syntetiikkaan, Sigma-Aldrich®

Toimittaja: Merck
Vaara


M-Clarity™ quality level = MQ200

8.21943.0100 8.21943.1000 8.21943.9020
8.21943.0100EA 38 EUR
8.21943.0100 8.21943.1000 8.21943.9020
2-Ethylhexyl phosphate (mono and di ester mixture) syntetiikkaan, Sigma-Aldrich®
2-Ethylhexyl phosphate (mono and di ester mixture)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00135884
CAS-numero: 12645-31-7
EINECS: 235-741-0
UN: 3265
ADR: 8,II

Order Now

Testitulosten erittely

Solubility (20 °C) insoluble
Boiling Point 180 °C (1013 hPa) decomposes
Flash Point 100 °C
Density 1.02 g/cm3 (20 °C)
pH 2.3 (10 g/l, H2O, 20 °C) (as an emulsion)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR