Isonicotinaldehyde oxime

Toimittaja: Merck
Varoitus


M-Clarity™ quality level = MQ200

8.20714.0050
8.20714.0050EA 229 EUR
8.20714.0050
Isonicotinaldehyde oxime
Isonicotinaldehyde oxime
Kaava: C₆H₆N₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006446
CAS-numero: 696-54-8
EINECS: 211-796-6

Order Now

Testitulosten erittely

Solubility 10 g/l (20 °C)
Melting Point 130 - 133 °C
Molar Mass 122.12 g/mol
Bulk Density 610 kg/m3
Vapor Pressure  - 0.01 hPa (50 °C)
Flash Point 110 °C DIN 51758
pH 6.3 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR