(-)-Di-p-toluoyl-L-tartaric acid for synthesis, Sigma-Aldrich®

Toimittaja: Merck

Synonyms: Di-p-toluoyl-L-tartaric acid, (-)-O,O'-Di-p-toluoyl-L-tartaric acid, (2R,3R)-(-)-Di-O-4-Toluoyl-L-Tartaric Acid

M-Clarity™ quality level = MQ200

8.18725.0025 8.18725.0100
8.18725.0025EA 112 EUR
8.18725.0025 8.18725.0100
(-)-Di-p-toluoyl-L-tartaric acid for synthesis, Sigma-Aldrich®
(-)-Di-p-toluoyl-L-tartaric acid
Kaava: C₂₀H₁₈O₈
MDL numero: MFCD00064440
CAS-numero: 32634-66-5
EINECS: 251-131-7

Order Now

Testitulosten erittely

Solubility (20 °C) slightly soluble
Melting Point 165 - 171 °C
Molar Mass 386.36 g/mol
Bulk Density 500 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR