Sitruunahappo for synthesis, Sigma-Aldrich®

Toimittaja: Merck
Varoitus

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

M-Clarity™ quality level = MQ200

8.18707.1000 8.18707.5000
8.18707.1000EA 63.7 EUR
8.18707.1000 8.18707.5000
Sitruunahappo for synthesis, Sigma-Aldrich®
Sitruunahappo
Kaava: HOC(COOH)(CH₂COOH)₂
Kiehumispiste: 310 °C (1013 hPa)
Sulamispiste: 149…151 °C
Tiheys: 1,665 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011669
CAS-numero: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Solubility 1330 g/l (20 °C)
Melting Point 153 °C (decomposition)
Molar Mass 192.12 g/mol
Bulk Density 560 kg/m3
Boiling Point 200 °C (1013 hPa) (decomposition)
Vapor Pressure  - 0.1 hPa (20 °C)
Density 1.665 g/cm3 (18 °C)
pH 1.7 (100 g/l, H2O, 20 °C)
Ignition Point 345 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR