(S)-(-)-β-Citronellol for synthesis, Sigma-Aldrich®

Toimittaja: Merck
Varoitus

Synonyms: (S)-(-)-3,7-Dimethyl-6-octen-1-ol, (S)-(-)-ß-Citronellol, (S)-(-)-Citronellol, (-)-beta-Citronellol, (-)-β-Citronellol, (S)-3,7-Dimethyloct-6-en-1-ol, beta-Rhodinol

M-Clarity™ quality level = MQ200

8.18378.0025 8.18378.0010 8.18378.9025 8.18378.8000
8.18378.0025EA 245 EUR
8.18378.0025 8.18378.0010 8.18378.9025 8.18378.8000
(S)-(-)-β-Citronellol for synthesis, Sigma-Aldrich®
(S)-(-)-β-Citronellol
Kaava: C₁₀H₂₀O
Tiheys: 0,855 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00063214
CAS-numero: 7540-51-4
EINECS: 231-415-7
UN: 3082
ADR: 9,III

Order Now

Testitulosten erittely

Solubility (20 °C) practically insoluble
Molar Mass 156.27 g/mol
Boiling Point 109 - 111 °C (13 hPa)
Flash Point 101 °C DIN 51758
Refractive Index 1.4562 (20 °C, 589 nm)
Density 0.855 g/cm3 (20 °C)
Ignition Point 240 °C
Synonyms (S)-(-)-3,7-Dimethyl-6-octen-1-ol, ß-Rhodinol
Hill Formula C10H20O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR