Hydrogen peroxide-urea, Sigma-Aldrich®

Toimittaja: Merck
Vaara


M-Clarity™ quality level = MQ200

8.18356.0100 8.18356.0500 8.18356.9010 8.18356.9025
8.18356.0100EA 38.6 EUR
8.18356.0100 8.18356.0500 8.18356.9010 8.18356.9025
Hydrogen peroxide-urea, Sigma-Aldrich®
Hydrogen peroxide-urea
Kaava: CH₆N₂O₃
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013119
CAS-numero: 124-43-6
EINECS: 204-701-4
UN: 1511
ADR: 5.1 (8),III

Order Now

Testitulosten erittely

Solubility 500 g/l (20 °C)
Melting Point 75 - 85 °C (decomposition)
Molar Mass 94.07 g/mol
pH 6.3 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR