9-Aminoacridine hydrochloride monohydrate ≥97.0% syntetiikkaan, Sigma-Aldrich®

Toimittaja: Merck


M-Clarity™ quality level = MQ200

8.01112.0025 8.01112.0100 8.01112.0002
8.01112.0025EA 125 EUR
8.01112.0025 8.01112.0100 8.01112.0002
9-Aminoacridine hydrochloride monohydrate ≥97.0% syntetiikkaan, Sigma-Aldrich®
9-Aminoacridine hydrochloride monohydrate
Kaava: C₁₃H₁₁ClN₂·1H₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00150071
CAS-numero: 52417-22-8
EINECS: 205-145-5
UN: 2811
ADR: 6.1,III

Order Now

Testitulosten erittely

CAS 52417-22-8
Hill Formula C₁₃H₁₁ClN₂ * H₂O
Chemical formula C₁₃H₁₀N₂ *HCl* H₂O
Molar Mass 248.71 g/mol
Melting Point 240 - 244 °C
Solubility 3.3 g/l
Assay (argentometric, calc. on anhydrous substance) ≥ 97,0 %(m)
Loss on drying 6,0 - 8,0 %(m)
Identity (IR) conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR