Valeriaanahappo for synthesis, Sigma-Aldrich®

Toimittaja: Merck
Vaara

Synonyms: Pentanoic acid

M-Clarity™ quality level = MQ200

8.00821.0100 8.00821.0500 8.00821.1000
8.00821.0100EA 26.6 EUR
8.00821.0100 8.00821.0500 8.00821.1000
Valeriaanahappo for synthesis, Sigma-Aldrich®
Valeriaanahappo
Kaava: C₅H₁₀O₂
Kiehumispiste: 186 °C (1013 hPa)
Sulamispiste: –32 °C
Tiheys: 1,11 g/cm³ (20 °C)
Leimahduspiste: 89 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004413
CAS-numero: 109-52-4
EINECS: 203-677-2
UN: 3265
ADR: 8,I
Merck Index: 12,10042

Order Now

Testitulosten erittely

Solubility 40 g/l (20 °C)
Melting Point -32 °C
Molar Mass 102.13 g/mol
Boiling Point 186 °C (1013 hPa)
Vapor Pressure 0.3 hPa (20 °C)
Flash Point 86 °C
Refractive Index 1.4076 (20 °C, 589 nm)
Explosion Limit 1.8 - 7.3 %(V)
Density 0.938 g/cm3 (20 °C)
pH 2.7 (40 g/l, H2O, 20 °C)
Ignition Point 360 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR