Supelco® TraceCERT® Organic Reference Standard, Metoxuron
Toimittaja: Merck
79828-50MG
79828-50MGEA 75.6 EUR
79828-50MG
Supelco® TraceCERT® Organic Reference Standard, Metoxuron
Standardiaineet ja vertailumateriaalit Organic Standards
Sertifikaatit: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Varoitus: P264 - P270 - P273 - P301 + P312 - P391 - P501