J.T.Baker® BAKERBOND® PROchievA™, Affinity Chromatography Resin, Recombinant Protein A
Toimittaja: Avantor
7899-01EA 1882.09 EUR
7899-01 7899-02 7899-03 7899-04 7899-05
J.T.Baker® BAKERBOND® PROchievA™, Affinity Chromatography Resin, Recombinant Protein A
Proteiininpuhdistusjärjestelmät Protein Purification Resins
The J.T.Baker® BAKERBOND® PROchievA™ protein A resin offers superior dynamic binding capacity with excellent alkaline stability linked to Avantor's proprietary ligand.
The J.T.Baker® BAKERBOND™ PROchievA™ protein A resin is designed for high-performance in the critical affinity chromatography step used in the purification of mAbs, Fc fusion proteins, and other similar IgG-like molecules.
Designed and manufactured by Avantor to the high standards established for our J.T.Baker® brand chemicals, BAKERBOND™ PROchievA™ protein A resin can be used with other J.T.Baker® BAKERBOND™ resins to enhance the separation power of your chromatography process steps.
Tilaustiedot: 1 ml and 5 ml lab columns are also available under complementary products.