Immobilised Ficin
Toimittaja: G Bioscienes
786-793
786-793EA 543 EUR
786-793
Immobilised Ficin
Proteiinien puhdistusjärjestelmät Proteiinien uuttoreagenssit (Extraction)
Ficin (or Ficain) is a cysteine protease enzyme (EC 3.4.22.3) isolated from fig latex is that has the endopeptidase activity to cleave Immunoglobulin G (IgG) molecules in the hinge region.
  • Generate Fab and F(ab‘)₂ fragments
  • For digestion of mouse IgG₁
  • Eliminates contamination by Ficin