Kuoppalevyt, Well-Coated™
Toimittaja: G-Biosciences
786-731 786-733 786-777 786-764 786-765 786-766 786-767 786-768 786-769 786-770 786-771 786-772 786-773 786-774 786-776 786-761 786-778 786-779 786-780 786-781 786-775 786-749 786-737 786-739 786-741 786-743 786-745 786-763 786-762 786-751 786-753 786-755 786-756 786-757 786-758 786-759 786-760 786-747 786-735
786-731EA 560 EUR
786-731 786-733 786-777 786-764 786-765 786-766 786-767 786-768 786-769 786-770 786-771 786-772 786-773 786-774 786-776 786-761 786-778 786-779 786-780 786-781 786-775 786-749 786-737 786-739 786-741 786-743 786-745 786-763 786-762 786-751 786-753 786-755 786-756 786-757 786-758 786-759 786-760 786-747 786-735
Kuoppalevyt, Well-Coated™
Kuoppalevyt Mikrolevyt immunologiaan
Pre-coated, blocked microtitre plates for screening and rapid assays.
  • Available in clear, black or white for colorimetric, chemiluminescent and fluorescent detection systems
  • 8 well strip format, supplied in convenient 96 well holder
  • Pre-blocked to save time
Protein A, G or A/G
Binds the constant (Fc) region of immunoglobulins ensuring that the antigen binding domain of the antibody is orientated away from the plate, offering maximum exposure of the binding site.
Protein L
Binds the kappa light chains of immunoglobulins without interfering with theMore Product Information