DNAOUT™ DNA Removal Solution
Toimittaja: G Bioscienes
786-74 786-75
786-74EA 82.2 EUR
786-74 786-75
DNAOUT™ DNA Removal Solution
Puhdistusaineet Dekontaminaatioliuokset nukleiinihapoille
DNAOUT™ is a DNA removal solution for the rapid decontamination of work areas from contaminating DNA. Simply spray the work area and wait 10-15 seconds and then rinse with Molecular Grade Water.
  • Ready-to-use with convenient spray bottle and refill bottles available
  • Remove contaminating DNA prior to sensitive work, including PCR
  • Removes amplicon, plasmid and genomic DNA
  • Fast acting (10-15 seconds)
  • Ideal for cleaning centrifuge tubes, reaction tubes, PCR machine surface, pipettes, lab benches, lab equipment and more