Tween® 80

Toimittaja: G Bioscienes

786-519 786-520
786-519EA 185 EUR
786-519 786-520
Tween® 80
Tween® 80

Tween® 80 for solubilising membrane proteins during isolation of membrane-protein complexes.

Sulamispiste: -21
MDL numero: MFCD00082107
CAS-numero: 9005-65-6
Merck Index: 13,07664
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bde3ba50379784286f232046e51f1c2e5aef0684&vid=764b1ae6c7de631c0dca755bd341e627e61103f4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR