SAM Methyltransferase assay, SAM510™
Toimittaja: G Bioscienes
786-430
786-430EA 1370 EUR
786-430
SAM Methyltransferase assay, SAM510™
Bio-assayt Entsymaattiset pikatestit
SAM510™ assay is a continuous enzyme coupled assay that can continuously monitor SAM-dependent methyltransferases without the use of radioactive labels or endpoint measurements.
  • Detection of protein methylation or screening methylation inhibitors
  • Continuous enzyme coupled assay for kinetic studies
  • Colorimetric, non radioactive assay
  • Supplied with all reagents, including positive control
  • Adaptable for enzymes that generate S-adenosylhomocysteine or 5’-methylthioadenosine
The assay can be adapted to be used with any SAM dependent methyltransferase or an enzyme reaction that produces 5-adenosylhomocysteine or 5’-methylthioadenosine, due to the specificity of adenosylhomocysteine nucleosidase.