Genomic DNA isolation and purification kits, GET™ DNA Template
Toimittaja: G Bioscienes
786-353 786-354
786-353EA 404 EUR
786-353 786-354
Genomic DNA isolation and purification kits, GET™ DNA Template
Nukleiinihapporeagenssit Nucleic Acid Purification Kits and Reagents
GET™ DNA Template is a spin column format kit suitable for the preparation of DNA templates from blood, cells, fungal and animal tissue samples. The isolated genomic DNA is suitable for PCR and other downstream applications.
  • Spin column format
  • Purifies mammalian genomic DNA free from impurities
  • Yield is ~1 to 40 µg DNA per preparation