CytoScan™ Cell proliferation assay kit, WST-1
Toimittaja: G Bioscienes
786-212
786-212EA 599 EUR
786-212
CytoScan™ Cell proliferation assay kit, WST-1
Bio-assayt Solujen testausanalyysit
CytoScan™ WST-1 Cell Proliferation Assay principle is based upon the reduction of the tetrazolium salt WST-1 to formazan by cellular dehydrogenases. The generation of the dark yellow coloured formazan is measured at 420 to 480 nm (optimal at 440 nm) and is directly correlated to cell number.
  • Sensitive and accurate assay
  • Colorimetric assay
  • No washing, harvesting or solubilization required
  • For the detection of cell toxicity, death, viability or proliferation
  • Ideal for high throughput screening