Magnetic beads, Sera-Mag SpeedBeads Neutravidin™-coated magnetic particles
Toimittaja: Cytiva
78152104011150 78152104010150 78152104010350
78152104011150.EA 304 EUR
78152104011150. 78152104010150. 78152104010350.
Magnetic beads, Sera-Mag SpeedBeads Neutravidin™-coated magnetic particles
Helmet ja partikkelit Magnetic Beads
Sera-Mag SpeedBeads Neutravidin™-Coated Magnetic Particles exhibit high affinity for biotinylated ligands such as proteins, nucleic acids, and peptides.
  • Even coating of Neutravidin™ protein provides reliable results
  • Low non-specific binding for fast and clean purification
  • Fast reaction kinetics increases throughput and precision, and also enables faster movement through viscous solutions
  • Unique cauliflower-like surface provides increased area for binding reactions compared to smooth surface particles
  • Uniform, nominal 1 μm Ø provides excellent lot-to-lot