Engine models
Toimittaja: CORNELSEN EXPERIMENTA
49206 49205 49204 49203 49202 49201
765-0176EA 0 EUR
765-0176 765-0175 765-0174 765-0173 765-0172 765-0171
Engine models
Opetusmateriaalit Physics Educational Materials Mechanics Learning Activities
Intended for use on overhead projectors for demonstration of function and principle for different types of motors. Made of strong, transparent plastic. Equipped with suction feet.
  • Dimensions: 250 × 250 mm