Spinal cord with nerve roots
Toimittaja: SOMMER MARCUS
BS32/37
763-0441EA 0 EUR
763-0441
Spinal cord with nerve roots
Mallit Ihmisbiologia
Nerve threads start from the gray spinal cord substance, cross through the white substance and gather into bundles of nerves. Magnification about 5X. A cross section of the spinal cord in about 10X magnification is mounted on the same plate.
  • The spinal ganglia are clearly visible in both models