Storage Boxes
Toimittaja: GRATNELL'S
F2TRAY F3 P5 F/1-G F/1-R F/1-B F/1-Y F/2-G F/2-R F/2-B F/2-Y F/3-G F/3-R F/3-B F/3-Y TCF/1
763-0313EA 41.3 EUR
763-0313 763-0314 763-0316 763-0301 763-0302 763-0300 763-0303 763-0305 763-0306 763-0304 763-0307 763-0309 763-0310 763-0308 763-0311 765-0964
Storage Boxes
Laatikot Säilytyslaatikot
Polypropylene storage trays. The material is fire-resistant and resistant to most chemicals in the school environment. Stackable.
  • BSI tested and passed for heavy educational use
  • All trays fit standard school furniture
  • Lids and inserts also available
  • Fully recyclable