Anti-CD8A Rat Monoclonal Antibody (violetFluor® 450) [clone: 53-6.7]
Toimittaja: Tonbo Biosciences
75-0081-U100 75-0081-U025
75-0081-U100EA 151 EUR
75-0081-U100 75-0081-U025
Anti-CD8A Rat Monoclonal Antibody (violetFluor® 450) [clone: 53-6.7]
Vasta-aineet
The 53-6.7 antibody reacts with the 32-34 kDa alpha subunit of mouse CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor in antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development,More Product Information