J.T.Baker® BAKERBOND spe™ C18 PolarPlus®, SPE Columns
Toimittaja: Avantor
7466-07EA 240.31 EUR
7466-07 7466-08 7466-06 7466-04 7466-01 7466-03 7466-10
J.T.Baker® BAKERBOND spe™ C18 PolarPlus®, SPE Columns
Kromatografiakolonnit
BAKERBOND spe™ columns are disposable PP or glass columns pre-packed with high capacity sorbents contained between two 20 µm PE frits. BAKERBOND spe™ C18 PolarPlus® columns are used when BAKERBOND spe™ C18 yields low recoveries of very polar (that is water soluble) compounds.
  • Silica based
  • Extraction of compounds from sample solution volumes ranging from a few hundred µl up to 1 L
  • Processing by vacuum, positive pressure or centrifugation
  • Yield up to 10 times higher recovery of polar compounds than BAKERBOND spe™ C18
General applications: General drugs (More Product Information