J.T.Baker® BAKERBOND® Q2100, New Generation UHPLC and HPLC Columns
Toimittaja: Avantor
7358-14EA 581 EUR
7358-14 7395-52 7342-52 7351-52 7395-12 7342-12 7351-12 7405-52 7354-52 7358-52 7373-52 7405-32 7354-32 7358-32 7373-32 7405-42 7354-42 7358-42 7373-42 7405-54 7354-54 7358-54 7373-54 7405-34 7354-34 7358-34 7373-34 7405-14 7354-14 7375-42 7411-22 7378-22 7382-22 7375-22 7411-14 7378-14 7382-14 7375-14 7411-24 7378-24 7382-24 7375-24 7373-14 7411-42 7378-42 7382-42 7411-2H 7411-4H 7378-2H 7378-4H 7382-2H 7382-4H 7375-2H 7375-4H 7395-2U 7342-2U 7351-2U 7354-24 7354-44 7358-24 7358-44 7373-24 7373-44 7375-44 7378-44 7382-44 7405-24 7405-44 7411-44 7405-64 7411-64 7354-64 7378-64 7382-26
J.T.Baker® BAKERBOND® Q2100, New Generation UHPLC and HPLC Columns
Kromatografiakolonnit
BAKERBOND® Q2100 LC phases have been specially designed to solve the challenges of most routine samples. They can also be a good starting point for new method development projects. With options of four widely used chemistries and three particle sizes BAKERBOND® Q2100 can be a useful addition to your analytical toolkit.
  • High efficiency UHPLC/HPLC columns
  • Based on high purity spherical silica
  • C18, C8, phenyl-hexyl and silica phases
  • Available with 1,8, 3, 5 µm particle sizes