Deepwell Plates 384-Well, 200 µl
Toimittaja: EPPENDORF
0030521.102 0030522.109 0030525.108 0030526.104 0030113.551
737-2549EA 461 EUR
737-2549 737-2554 737-0210 737-0211 737-0253
Deepwell Plates 384-Well, 200 µl
Kuoppalevyt Syväkuoppalevyt
Eppendorf Deepwell plates 384-well, 200 µl offer efficient sample handling with features like OptiTrack® matrix for rapid identification and g-Safe® technology for stability during centrifugation. SafeCode variants provide secure three-level coding, including error correction for reliable identification, ensuring flexibility and accuracy in experimental workflows.
  • OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
  • RecoverMax® well design: Optimized well geometry for maximum sample