Eppendorf® DNA LoBind Plates
Toimittaja: EPPENDORF
0030503.104 0030503.147 0030503.201 0030503.244 0030507.100 0030507.207 0030527.100 0030523.105 0030623.304 0030603.303
737-2530EA 452 EUR
737-2530 737-2534 737-2535 737-2539 737-0206 737-0207 737-0212 737-2559 737-0162 737-0214
Eppendorf® DNA LoBind Plates
Kuoppalevyt Syväkuoppalevyt
DNA LoBind Plates maximise sample recovery of nucleic acids by significantly reducing sample-to-surface binding. These plates are the ideal solution for sample preparation and long-term storage.
  • Eppendorf LoBind material ensures optimised sample recovery for improved assay results
  • Free of surface coating (e.g. silicone) to minimise the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • High-contrast Unique OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors