Automation Tips for Tecan Fluent and Evo Workstations
Toimittaja: FLUOTICS
LHA-1000.F LHA-1000.ST LHA-1000CB.F LHA-1000CB.ST LHA-1000CBR.F LHA-1000CBR.ST LHA-1000R.F LHA-1000R.ST LHA-10CB.F LHA-10CB.ST LHA-10CBR.F LHA-10CBR.ST LHA-200.F LHA-200.ST LHA-200CB.F LHA-200CB.ST LHA-200CBR.F LHA-200CBR.ST LHA-200R.F LHA-200R.ST LHA-50.F LHA-50.ST LHA-50CB.F LHA-50CB.ST LHA-50CBR.F LHA-50CBR.ST LHA-50R.F LHA-50R.ST LHAB-01 LHAB-02 LHAB-1000CBR.F LHAB-1000CBRST LHAB-1000R.F LHAB-1000R.ST LHAB-10CBR.F LHAB-10CBR.ST LHAB-200CBR.F LHAB-200CBR.ST LHAB-200R.F LHAB-200R.ST LHAB-50CBR.F LHAB-50CBR.ST LHAB-50R.F LHAB-50R.ST MCA-200XL.F MCA-200XL.NS MCA-200XL.ST MCA-50.F MCA-50.NS MCA-50.ST MCA-500.F MCA-500.NS MCA-500.ST
736-1008EA 601 EUR
736-1008 736-1009 736-1010 736-1011 736-1012 736-1013 736-1014 736-1015 736-1016 736-1017 736-1018 736-1019 736-1020 736-1021 736-1022 736-1023 736-1024 736-1025 736-1026 736-1027 736-1028 736-1029 736-1030 736-1031 736-1032 736-1033 736-1034 736-1035 736-1036 736-1037 736-1038 736-1039 736-1040 736-1041 736-1042 736-1043 736-1044 736-1045 736-1046 736-1047 736-1048 736-1049 736-1050 736-1051 736-1052 736-1053 736-1054 736-1055 736-1056 736-1057 736-1058 736-1059 736-1060
Automation Tips for Tecan Fluent and Evo Workstations
Pipetinkärjet
Fluotics manufactures a complete selection of automation tips compatible with Tecan fluent and Evo workstations. These automation tips are directly interchangeable with their Tecan automation tip counterpart, with no change to the labware definitions required.
  • Certified RNase-/DNase-free and nonpyrogenic
  • Produced from virgin, high-grade polypropylene
  • Available in non sterile, sterile, and barrier filter formats
Compatible with Tecan fluent and Evo workstations with MCA and LiHA/FCA heads. Ultra polishing techniques ensure low liquid retention withoutMore Product Information