Automation Tips for Agilent® Bravo and VPrep® Workstations
Toimittaja: FLUOTICS
AGI-10.NS AGI-10.ST AGI-250.F AGI-250.F/A AGI-250.NS AGI-250.NS/A AGI-250.ST AGI-250.ST/A AGI-30.F AGI-30.F/A AGI-30.NS AGI-30.NS/A AGI-30.ST AGI-30.ST/A AGI-70.F AGI-70.NS AGI-70.ST AGI-70WB.F AGI-70WB.NS AGI-70WB.ST
736-0932EA 2650 EUR
736-0932 736-0933 736-0934 736-0935 736-0936 736-0937 736-0938 736-0939 736-0940 736-0941 736-0942 736-0943 736-0944 736-0945 736-0946 736-0947 736-0948 736-0949 736-0950 736-0951
Automation Tips for Agilent® Bravo and VPrep® Workstations
Pipetinkärjet
Fluotics manufactures a complete selection of automation tips compatible with the Agilent™ Bravo and VPrep. The automation tips are directly interchangeable with their Agilent robotic tip counterpart with no change to the labware definitions required.
  • Certified RNase-/DNase-free and nonpyrogenic
  • Produced from virgin, high grade polypropylene
  • Available in non-sterile, sterile and barrier filter formats
Compatible with Agilent™ VPrep® or Bravo® 96 or 384 ST series (Short tip, type II or III) multi-channels heads. Ultra polishing techniques ensure lowMore Product Information