Nucleic acid and protein purification system, KingFisher™ Duo Prime
Toimittaja: Thermo Fisher Scientific
5400110
736-0901EA 25680 EUR
736-0901
Nucleic acid and protein purification system, KingFisher™ Duo Prime
Automaattiset puhdistusjärjestelmät nukleiinihapoille
KingFisher™ Duo Prime magnetic particle processor is a compact, flexible purification system that is ideal for laboratories requiring nucleic acid purification, and is easily modified for use in proteomics and cell isolation. It provides excellent reproducibility with minimal hands-on time, and has a small footprint in the laboratory. Either small-volume (10 to 50 µl) or large-volume (300 to 500 µl) samples can be processed on the KingFisher™ Duo Prime, due to its interchangeable magnetic rod design.
  • Easy installation (ready to run in 10 minutes or less)
  • Flexible configuration includes