Kuoppalevypesuri, Wellwash™
Toimittaja: THERMO LABSYSTEMS LIFE SCIENCE
5165000 5165040
736-0422EA 6140 EUR
736-0422 736-3008
Kuoppalevypesuri, Wellwash™
Kuoppalevypesurit
Wash 96-well plates securely with the Thermo Scientific™ Wellwash™ Microplate Washer, an easy-to-use, reliable microplate-strip washer for routine ELISA applications. The Wellwash features a graphical user interface, local language versions and USB port for optimal ease-of-use and convenience.
  • Large color clear display and keypad for easy, convenient use
  • User interface is logic and intuitive for quick protocols development
  • Easy-to-use software requires minimal training for fast startup
  • Unpressurized bottles are safe and secure to use
  • Liquid level sensors