MultiScreen® Vacuum Manifold 96 Well
Toimittaja: Merck
MAVM0960R
736-0190EA 2140 EUR
736-0190
MultiScreen® Vacuum Manifold 96 Well
Manifoldit
96-well plate Vacuum Manifold for vaccum filtration.
Components: EPDM rubber gasket, EPDM rubber seal, nylon well ring (standard), Tygon® tubing (FEP-lined), brass socket (for vacuum control valve), high-density polyethylene base, high-density polyethylene well ring (deep), polypropylene valve (on/off), stainless steel grid (for support), steel case (for vacuum control valve), steel gauge (for vacuum).