MultiScreen Filter Plates with Hydrophilic Durapore® PVDF Membrane
Toimittaja: Merck
MAHVN4550 MADVN6550 MAGVS2210 MAHVN4510 MABVN1250 MABVN0B50
736-0186EA 1600 EUR
736-0186 736-0177 736-0181 736-0185 736-0175 MILFMABVN0B50
MultiScreen Filter Plates with Hydrophilic Durapore® PVDF Membrane
Kuoppalevyt Suodatinlevyt
MultiScreen HTS filtration plates are used to filter samples and perform entire procedures, from cell growth to scintillation counting within the same plate.
  • Hydrophilic Durapore® polyvinylidene fluoride (PVDF) membrane
The plates come in many membrane types, pore sizes, and plate materials. The automation-compatible design allows for easy manipulation with a variety of robotics systems and makes barcoding possible on all four sides. Filtration is accomplished either by vacuum using the MultiScreen HTS Vacuum Manifold, or by centrifugation.