Soluviljelypullot, Corning®
Toimittaja: Corning
430372 430639 430720 430824 430825 431079 431080 431081 431082 431085 431306 430168 430823 3292 3290 3289 3291 3293 431328 3814 431463 431465 431466 430641U 430720U 430725U 431464U
734-1706EA 723 EUR
734-1706 734-1712 734-1715 734-1718 734-1719 734-1722 734-1723 734-1724 734-1725 734-1726 734-1733 734-1700 734-1717 734-0093 734-0091 734-0090 734-0092 734-0094 734-1214 734-4139 734-2272 734-2274 734-2275 734-2705 734-2706 734-2707 734-2708
Soluviljelypullot, Corning®
Kolvit ja erlenmeyerit Soluviljelypullot
Manufactured from optically clear virgin polystyrene.
  • Printed with lot numbers for ease in traceability
  • 100% integrity tested
  • Sterilised by gamma irradiation and certified non pyrogenic
  • Canted models have a skirt to prevent tipping
Non-treated: Flasks are designed for use in applications where cell attachment is not desired.
TC treated: For optimal cell attachment and growth.
Ultra low attachment surface: Features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation,More Product Information