96-well solid black and white polystyrene microplates, Corning®
Toimittaja: Corning
3362 3600 3650 3688 3693 3694 3792 3912 3915 3916 3917 3922 3925 3605 3686 3642 3990 3993 3991 3875 4591 3789A
734-1549EA 634 EUR
734-1549 732-5515 732-5522 734-1623 734-1625 734-1626 734-1643 734-1662 734-1663 734-1664 734-1665 734-1666 734-1667 732-5517 734-1622 734-4117 732-5596 734-4171 734-4169 734-4153 CORN4591 CORN3789A
96-well solid black and white polystyrene microplates, Corning®
Kuoppalevyt Soluviljelylevyt, monikuoppaiset
These 96-well solid black and white polystyrene microplates are designed to reduce well-to-well cross-talk.
White microplates enhance luminescent signals and have low background luminescence.
Black microplates have low background fluorescence and minimise light scattering.